Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810009A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025463.3 | NP_079739.1 | 4 | 402 | ||
GenBank mRNA | AK007390.1 | - | 4 | 383 | ||
BC047099.1 | - | 4 | 381 | 5253822 | ||
BC058702.1 | - | 4 | 365 | 6822952 |
Gene Symbol: Lbhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286103.1 | NP_001273032.1 | 6 | 898 |