Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025824.3 | NP_080100.1 | 3 | 176 | ||
GenBank mRNA | AK004784.2 | - | 3 | 176 | ||
AK013980.1 | - | 3 | 170 | |||
AK029030.1 | - | 3 | 187 | |||
AK076086.1 | - | 3 | 177 | |||
AK076560.1 | - | 3 | 177 | |||
AK087980.1 | - | 3 | 164 | |||
AK088322.1 | - | 3 | 184 | |||
AK133973.1 | - | 3 | 177 | |||
AK151919.1 | - | 3 | 164 | |||
AK152939.1 | - | 3 | 164 | |||
AK153538.1 | - | 3 | 165 | |||
AK161192.1 | - | 3 | 174 | |||
AK166247.1 | - | 3 | 181 | |||
AK167056.1 | - | 3 | 161 | |||
AK167074.1 | - | 3 | 148 | |||
AK167514.1 | - | 3 | 136 | |||
AK167982.1 | - | 3 | 182 | |||
AK168158.1 | - | 3 | 188 | |||
AK192968.1 | - | 3 | 141 | |||
AK195624.1 | - | 3 | 141 | |||
AK201364.1 | - | 232 | ||||
AK205016.1 | - | 3 | 171 | |||
AK205108.1 | - | 3 | 141 | |||
AK212098.1 | - | 3 | 141 | |||
BC005466.1 | - | 3 | 185 | 3601190 | ||
BC028865.1 | - | 3 | 164 | 4910833 |