Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026155.3 | NP_080431.1 | 3 | 299 | ||
GenBank mRNA | AK002818.1 | - | 3 | 300 | ||
AK029353.1 | - | 3 | 300 | |||
AK076235.1 | - | 3 | 332 | |||
AK088555.1 | - | 3 | 304 | |||
AK150255.1 | - | 3 | 293 | |||
AK192841.1 | - | 3 | 220 | |||
AK194871.1 | - | 3 | 220 | |||
AK199053.1 | - | 3 | 220 | |||
AK206034.1 | - | 3 | 220 | |||
BC011111.1 | - | 3 | 306 | 3965897 | ||
BC020335.1 | - | 3 | 271 | |||
CT010172.1 | - | 3 | 283 |