Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Saraf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026432.3 | NP_080708.3 | 2 | 415 | ||
GenBank mRNA | AK007787.1 | - | 2 | 342 | ||
AK075744.1 | - | 2 | 306 | |||
AK077118.1 | - | 2 | 402 | |||
AK088728.1 | - | 2 | 298 | |||
AK093942.1 | - | 2 | 390 | |||
AK163218.1 | - | 2 | 348 | |||
AK197284.1 | - | 1 | 32 | |||
AK207744.1 | - | 1 | 32 | |||
AK213493.1 | - | 1 | 32 | |||
BC013497.1 | - | 2 | 356 | 4222385 | ||
BC022616.1 | - | 2 | 393 | 4187981 | ||
BC024888.1 | - | 2 | 351 | 3988822 |