Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027456.1 | NP_081732.1 | 3 | 189 | ||
NM_030184.2 | NP_084460.1 | 3 | 224 | |||
GenBank mRNA | AK014450.1 | - | 3 | 194 | ||
AK017564.1 | - | 3 | 190 | |||
AK019512.1 | - | 3 | 219 | |||
AK019600.1 | - | 3 | 213 | |||
AK030307.1 | - | 3 | 221 | |||
AK036083.1 | - | 3 | 195 | |||
AK039800.1 | - | 3 | 197 | |||
AK045699.1 | - | 3 | 188 | |||
AK046693.1 | - | 3 | 207 | |||
AK048064.1 | - | 3 | 203 | |||
AK140598.1 | - | 3 | 220 | |||
AK161393.1 | - | 3 | 204 | |||
AK164041.1 | - | 3 | 224 | |||
BC043101.1 | - | 3 | 176 | 6406321 |