Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCCA SIRNA Assays ›
Gene Symbol:
PCCA
Gene Name:
propionyl-CoA carboxylase alpha subunit
Gene Aliases:
-
Chromosome Location:
Chr.13: 100089015 - 100530437 on Build GRCh38
UniGene ID:
Hs.733632
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
8192
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCCA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000282.3
NP_000273.2
6
538
NM_001127692.2
NP_001121164.1
5
460
NM_001178004.1
NP_001171475.1
6
538
XM_005254059.2
XP_005254116.1
6
533
XM_011521093.1
XP_011519395.1
6
533
XM_017020605.1
XP_016876094.1
6
533
XM_017020606.1
XP_016876095.1
5
384
XM_017020607.1
XP_016876096.1
6
349
XM_017020608.1
XP_016876097.1
6
533
XM_017020609.1
XP_016876098.1
6
446
XM_017020610.1
XP_016876099.1
4
800
XM_017020611.1
XP_016876100.1
6
533
XM_017020612.1
XP_016876101.1
6
533
XM_017020613.1
XP_016876102.1
6
533
XM_017020614.1
XP_016876103.1
5
384
XM_017020615.1
XP_016876104.1
6
533
XM_017020616.1
XP_016876105.1
6
533
XM_017020617.1
XP_016876106.1
3
196
XM_017020618.1
XP_016876107.1
3
196
GenBank mRNA
AF385926.1
-
6
470
AK055982.1
-
4
800
AK225288.1
-
6
438
AK225299.1
-
6
436
AK296771.1
-
5
358
AK298318.1
-
6
460
AK315503.1
-
6
436
BC000140.2
-
6
410
3048998
DA781564.1
-
5
358
X14608.1
-
6
405
Z36837.1
-
2
132
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biotin metabolic process
short-chain fatty acid catabolic process
biotin carboxylase activity
propionyl-CoA carboxylase activity
ATP binding
biotin binding
enzyme binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0