Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810403A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028814.3 | NP_083090.2 | 2 | 290 | ||
GenBank mRNA | AK012969.1 | - | 2 | 267 | ||
AK020778.1 | - | 2 | 241 | |||
AK038617.1 | - | 2 | 271 | |||
AK083018.1 | - | 2 | 251 | |||
AK164337.1 | - | 2 | 251 | |||
AK170490.1 | - | 2 | 285 | |||
AK173057.1 | - | 2 | 254 | |||
BC006621.1 | - | 2 | 265 | 3158104 | ||
BY097934.1 | - | 2 | 291 | |||
CX562176.1 | - | 2 | 256 |