Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810403A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028814.3 | NP_083090.2 | 14 | 1796 | ||
GenBank mRNA | AK012969.1 | - | 14 | 1773 | ||
AK170490.1 | - | 14 | 1791 | |||
AK173057.1 | - | 13 | 2508 | |||
BC006621.1 | - | 14 | 1771 | 3158104 |