Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054055.2 | NP_473396.1 | 1, 2 | 155 | ||
GenBank mRNA | AF306491.1 | - | 1, 2 | 154 | ||
AK085326.1 | - | 1, 2 | 153 | |||
AK086775.1 | - | 1, 2 | 157 | |||
AK087518.1 | - | 1, 2 | 153 | |||
AK128982.1 | - | 1, 2 | 152 | |||
AK138344.1 | - | 1, 2 | 153 | |||
AK157881.1 | - | 1, 2 | 124 | |||
AK165355.1 | - | 1, 2 | 153 | |||
BC026803.1 | - | 1, 2 | 141 | 4238769 | ||
BC028882.1 | - | 1, 2 | 139 |