Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chtf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145412.3 | NP_663387.3 | 4 | 1286 | ||
GenBank mRNA | AK030924.1 | - | 4 | 1285 | ||
AK088257.1 | - | 4 | 1295 | |||
AK165622.1 | - | 4 | 1268 | |||
AK165993.1 | - | 2 | 1112 | |||
BC023107.1 | - | 2 | 1058 | 3966700 |