Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145575.3 | NP_663550.1 | 2 | 356 | ||
GenBank mRNA | AK050642.1 | - | 4 | 426 | ||
AK160593.1 | - | 2 | 268 | |||
BC019435.1 | - | 2 | 315 | 5148205 | ||
BC055908.1 | - | 4 | 429 | |||
BY308240.1 | - | 2 | 357 |