Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153415.3 | NP_700464.2 | 3, 4 | 928 | ||
GenBank mRNA | AF246235.1 | - | 3, 4 | 532 | ||
AK043573.1 | - | 3, 4 | 629 | |||
AK043713.1 | - | 3, 4 | 470 | |||
AK050915.1 | - | 3, 4 | 497 | |||
AK076813.1 | - | 3, 4 | 930 | |||
AK085703.1 | - | 3, 4 | 997 | |||
AY090481.1 | - | 3, 4 | 928 | |||
AY090482.1 | - | 3, 4 | 528 | |||
BC052045.1 | - | 3, 4 | 500 | 6402328 | ||
BC062965.1 | - | 3, 4 | 500 | |||
BC079589.1 | - | 3, 4 | 498 |