Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153516.2 | NP_705736.1 | 2 | 269 | ||
GenBank mRNA | AK047223.1 | - | 2 | 271 | ||
AK088417.1 | - | 2 | 284 | |||
AK132774.1 | - | 2 | 278 | |||
AK143457.1 | - | 2 | 282 | |||
AK148007.1 | - | 2 | 288 | |||
AK165068.1 | - | 2 | 262 | |||
AK168833.1 | - | 2 | 259 | |||
AK171256.1 | - | 2 | 289 | |||
AK178476.1 | - | 1 | 42 | |||
AK179659.1 | - | 1 | 42 | |||
BC027307.1 | - | 2 | 159 | 4014844 | ||
BC029016.1 | - | 2 | 269 | 5358785 |