Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC029214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153557.1 | NP_705785.1 | 5 | 666 | ||
GenBank mRNA | AK046066.1 | - | 4 | 596 | ||
AK053101.1 | - | 3 | 278 | |||
BC029214.1 | - | 5 | 666 | 4161391 |