Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam210a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153794.4 | NP_722489.1 | 4 | 1600 | ||
GenBank mRNA | AK034980.1 | - | 4 | 1482 | ||
AK044238.1 | - | 4 | 1489 | |||
AK049567.1 | - | 4 | 1488 | |||
AK077093.1 | - | 4 | 1601 | |||
AK079137.1 | - | 1 | 723 | |||
AK142190.1 | - | 4 | 1472 | |||
AK147662.1 | - | 4 | 1474 | |||
AK159319.1 | - | 4 | 1449 | |||
AK162396.1 | - | 4 | 1486 | |||
BC038695.1 | - | 4 | 1368 | 1548421 | ||
BC095936.1 | - | 4 | 1449 | 3968496 |