Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ofcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172143.2 | NP_742155.2 | 3 | 157 | ||
GenBank mRNA | AF548110.1 | - | 4 | 441 | ||
AF548111.1 | - | 3 | 177 | |||
EU683438.1 | - | 3 | 157 | |||
EU683439.1 | - | 3 | 157 |