Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252584.2 | NP_001239513.1 | 4 | 502 | ||
NM_001252585.1 | NP_001239514.1 | 3 | 394 | |||
NM_001252587.2 | NP_001239516.1 | 5 | 514 | |||
NM_001281926.1 | NP_001268855.1 | 4 | 401 | |||
NM_001291158.1 | NP_001278087.1 | 5 | 2511 | |||
NM_027230.5 | NP_081506.3 | 5 | 589 | |||
GenBank mRNA | AB093284.2 | - | 5 | 2503 | ||
AK132427.1 | - | 3 | 396 | |||
AK147620.1 | - | 4 | 402 | |||
AK155864.1 | - | 4 | 307 | |||
BC023300.1 | - | 4 | 369 | |||
BC048186.1 | - | 5 | 396 | 6827541 | ||
BC063267.1 | - | 4 | 295 | |||
CA328656.1 | - | 1 | 18 | |||
CF537480.1 | - | 4 | 427 | |||
CN670238.1 | - | 5 | 329 | |||
CO428932.1 | - | 2 | 158 |