Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172749.4 | NP_766337.2 | 2 | 3018 | ||
GenBank mRNA | AK078520.1 | - | 2 | 2741 | ||
AK129112.1 | - | 1 | 1870 | |||
BC068300.1 | - | 1 | 2621 | 30105973 | ||
BC079907.1 | - | 2 | 3018 | 30647214 |