Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172803.2 | NP_766391.2 | 17 | 2005 | ||
GenBank mRNA | AK030389.1 | - | 17 | 2020 | ||
AK031713.1 | - | 17 | 1971 | |||
AK043666.1 | - | 17 | 2006 | |||
AK122353.1 | - | 17 | 1987 | |||
BC141138.1 | - | 17 | 1722 | 9055750 |