Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310035C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029349.1 | NP_083625.1 | 3, 4 | 927 | ||
NM_173187.3 | NP_775279.2 | 4, 5 | 999 | |||
GenBank mRNA | AK035724.1 | - | 4, 5 | 1001 | ||
AK040168.1 | - | 5, 6 | 635 | |||
AK129368.1 | - | 3, 4 | 900 | |||
AK148005.1 | - | 4, 5 | 979 | |||
BC060194.1 | - | 3, 4 | 172 | |||
BC117949.1 | - | 4, 5 | 933 | 40098078 | ||
BC117950.1 | - | 3, 4 | 865 | 40098082 |