Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9830107B12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177896.1 | NP_001171367.1 | 2 | 406 | ||
NM_177824.4 | NP_808492.1 | 2 | 406 | |||
GenBank mRNA | AK079398.1 | - | 2 | 367 | ||
EF646801.1 | - | 107 |