Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178388.2 | NP_848475.1 | 4 | 583 | ||
GenBank mRNA | AK008510.1 | - | 458 | |||
BC049695.1 | - | 4 | 583 | 6705996 | ||
BC132482.1 | - | 4 | 570 | 40130759 | ||
BC138196.1 | - | 4 | 570 | 8861215 |