Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDH18 SIRNA Assays ›
          Gene Symbol:
          CDH18
          Gene Name:
          cadherin 18
          Gene Aliases:
          CDH14, CDH14L, CDH24
          Chromosome Location:
          Chr.5: 19472930 - 20575902 on Build GRCh38
          UniGene ID:
          Hs.317632
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS101716
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDH18
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001167667.1 NP_001161139.1 13 2498
          NM_001291956.1 NP_001278885.1 15 3064
          NM_001291957.1 NP_001278886.1 13 2479
          NM_004934.3 NP_004925.1 13 2607
          XM_005248228.3 XP_005248285.1 13 2816
          XM_006714435.3 XP_006714498.1 12 2423
          XM_011513928.2 XP_011512230.1 11 2221
          XM_011513929.2 XP_011512231.1 10 2475
          XM_011513930.2 XP_011512232.1 13 2588
          XM_017008924.1 XP_016864413.1 14 2935
          XM_017008925.1 XP_016864414.1 14 2931
          XM_017008926.1 XP_016864415.1 14 2792
          XM_017008927.1 XP_016864416.1 14 2721
          XM_017008928.1 XP_016864417.1 15 2979
          XM_017008929.1 XP_016864418.1 12 2459
          XM_017008930.1 XP_016864419.1 12 2350
          XM_017008931.1 XP_016864420.1 13 2603
          XM_017008932.1 XP_016864421.1 14 2654
          XM_017008933.1 XP_016864422.1 14 2783
          XM_017008934.1 XP_016864423.1 14 2921
          XM_017008935.1 XP_016864424.1 14 2829
          XM_017008936.1 XP_016864425.1 13 2710
          XM_017008937.1 XP_016864426.1 13 2469
          XM_017008938.1 XP_016864427.1 13 2433
          XM_017008939.1 XP_016864428.1 14 2803
          XM_017008940.1 XP_016864429.1 13 2691
          XM_017008941.1 XP_016864430.1 13 2450
          GenBank mRNA AK289547.1 - 15 3062
          BC031051.1 - 13 2563 5278717
          CA425592.1 - 1 493
          U59325.1 - 12 2386

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          adherens junction organization
          calcium ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0