Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EYA3 SIRNA Assays ›
Gene Symbol:
EYA3
Gene Name:
EYA transcriptional coactivator and phosphatase 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 27970344 - 28088671 on Build GRCh38
UniGene ID:
Hs.185774
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS103456
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EYA3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282560.1
NP_001269489.1
14
1389
NM_001282561.1
NP_001269490.1
14
1389
NM_001282562.1
NP_001269491.1
14
1447
NM_001990.3
NP_001981.2
15
1527
NR_104214.1
16
1595
XM_006710449.3
XP_006710512.1
16
1563
XM_011540999.2
XP_011539301.1
16
1566
XM_011541001.2
XP_011539303.1
15
1524
XM_011541003.2
XP_011539305.1
15
1426
XM_011541004.2
XP_011539306.1
15
1423
XM_017000652.1
XP_016856141.1
16
1567
XM_017000653.1
XP_016856142.1
17
1687
XM_017000654.1
XP_016856143.1
15
1524
XM_017000655.1
XP_016856144.1
16
1680
XM_017000656.1
XP_016856145.1
15
1427
XM_017000657.1
XP_016856146.1
14
1419
XM_017000658.1
XP_016856147.1
14
1444
XM_017000659.1
XP_016856148.1
13
1303
GenBank mRNA
AJ007991.1
-
2
109
AK289805.1
-
15
1486
AK295745.1
-
14
1383
AK298129.1
-
14
1444
AK303664.1
-
14
1371
BC029500.1
-
16
1555
5272319
BC041667.1
-
14
1316
5743906
BQ018346.1
-
2
272
BX648945.1
-
15
1496
U81602.1
-
10
1126
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
visual perception
anatomical structure morphogenesis
response to ionizing radiation
histone dephosphorylation
peptidyl-tyrosine dephosphorylation
positive regulation of DNA repair
protein tyrosine phosphatase activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline