Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004486.4 | NP_004477.3 | 13 | 1007 | ||
XM_005251930.1 | XP_005251987.1 | 14 | 1106 | |||
XM_005251931.1 | XP_005251988.1 | 13 | 1025 | |||
XM_005251932.1 | XP_005251989.1 | 12 | 937 | |||
GenBank mRNA | AF248953.1 | - | 13 | 958 | ||
BC014188.2 | - | 1 | 24 | |||
BC069268.1 | - | 13 | 1007 | 6340972 |