Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ICA1 SIRNA Assays ›
Gene Symbol:
ICA1
Gene Name:
islet cell autoantigen 1
Gene Aliases:
ICA69, ICAp69
Chromosome Location:
Chr.7: 8113184 - 8262687 on Build GRCh38
UniGene ID:
Hs.487561
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS105158
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ICA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136020.2
NP_001129492.1
4
483
NM_001276478.1
NP_001263407.1
4
465
NM_004968.3
NP_004959.2
4
468
NM_022307.2
NP_071682.1
4
283
XM_005249735.3
XP_005249792.1
4
405
XM_011515345.1
XP_011513647.1
4
483
XM_011515346.1
XP_011513648.1
4
468
XM_011515347.2
XP_011513649.1
4
355
XM_011515348.1
XP_011513650.1
4
401
XM_011515349.1
XP_011513651.1
4
334
XM_011515350.1
XP_011513652.1
4
405
XM_011515351.1
XP_011513653.1
4
540
XM_011515352.1
XP_011513654.1
4
359
XM_011515353.2
XP_011513655.1
4
412
XM_017012116.1
XP_016867605.1
4
251
XM_017012117.1
XP_016867606.1
4
441
XM_017012118.1
XP_016867607.1
4
401
XM_017012119.1
XP_016867608.1
4
376
XM_017012120.1
XP_016867609.1
4
422
XM_017012121.1
XP_016867610.1
4
289
XM_017012122.1
XP_016867611.1
4
454
XM_017012123.1
XP_016867612.1
4
393
XM_017012124.1
XP_016867613.1
4
554
XM_017012125.1
XP_016867614.1
4
304
GenBank mRNA
AK292106.1
-
4
352
BC008640.2
-
4
400
3854722
DA076379.1
-
4
469
DB010744.1
-
4
452
DB040266.1
-
4
411
EU416199.1
-
3
216
L01100.1
-
4
394
L21181.1
-
3
292
U26591.1
-
4
283
U26592.1
-
4
340
U37183.1
-
2
182
U38260.1
-
4
385
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neurotransmitter transport
regulation of protein homodimerization activity
regulation of insulin secretion
molecular_function
protein binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0