Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ICA1 SIRNA Assays ›
          Gene Symbol:
          ICA1
          Gene Name:
          islet cell autoantigen 1
          Gene Aliases:
          ICA69, ICAp69
          Chromosome Location:
          Chr.7: 8113184 - 8262687 on Build GRCh38
          UniGene ID:
          Hs.487561
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS105159
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ICA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136020.2 NP_001129492.1 5, 6 649
          NM_001276478.1 NP_001263407.1 5, 6 631
          NM_004968.3 NP_004959.2 5, 6 634
          NM_022307.2 NP_071682.1 5, 6 449
          XM_005249735.3 XP_005249792.1 5, 6 571
          XM_011515345.1 XP_011513647.1 5, 6 649
          XM_011515346.1 XP_011513648.1 5, 6 634
          XM_011515347.2 XP_011513649.1 5, 6 521
          XM_011515348.1 XP_011513650.1 5, 6 567
          XM_011515349.1 XP_011513651.1 5, 6 500
          XM_011515350.1 XP_011513652.1 5, 6 571
          XM_011515351.1 XP_011513653.1 5, 6 706
          XM_011515352.1 XP_011513654.1 5, 6 525
          XM_011515353.2 XP_011513655.1 5, 6 578
          XM_011515354.1 XP_011513656.1 2, 3 162
          XM_017012116.1 XP_016867605.1 5, 6 417
          XM_017012117.1 XP_016867606.1 5, 6 607
          XM_017012118.1 XP_016867607.1 5, 6 567
          XM_017012119.1 XP_016867608.1 5, 6 542
          XM_017012120.1 XP_016867609.1 5, 6 588
          XM_017012121.1 XP_016867610.1 5, 6 455
          XM_017012122.1 XP_016867611.1 6, 7 834
          XM_017012123.1 XP_016867612.1 6, 7 773
          XM_017012124.1 XP_016867613.1 6, 7 934
          XM_017012125.1 XP_016867614.1 6, 7 684
          GenBank mRNA AK292106.1 - 5, 6 518
          BC008640.2 - 5, 6 566 3854722
          EU416199.1 - 4, 5 382
          L01100.1 - 5, 6 560
          L21181.1 - 5, 6 458
          U26591.1 - 5, 6 449
          U26592.1 - 6, 7 720
          U37183.1 - 3, 4 348
          U38260.1 - 5, 6 551

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter transport
          regulation of protein homodimerization activity
          regulation of insulin secretion
          molecular_function
          protein binding
          protein domain specific binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0