Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007245.2 | NP_001007246.1 | 6 | 945 | ||
NM_001197079.1 | NP_001184008.1 | 5 | 835 | |||
NM_001197080.1 | NP_001184009.1 | 5 | 670 | |||
NM_001550.3 | NP_001541.2 | 5 | 736 | |||
XM_011516142.2 | XP_011514444.1 | 5 | 731 | |||
XM_011516143.2 | XP_011514445.1 | 5 | 916 | |||
XM_017012151.1 | XP_016867640.1 | 5 | 983 | |||
GenBank mRNA | AK298894.1 | - | 5 | 476 | ||
AK308066.1 | - | 6 | 920 | |||
AK313211.1 | - | 6 | 945 | |||
AK315967.1 | - | 5 | 668 | |||
BC001272.1 | - | 5 | 689 | 3458309 | ||
BT019889.1 | - | 5 | 475 | |||
BT019890.1 | - | 5 | 475 |