Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MID1 SIRNA Assays ›
          Gene Symbol:
          MID1
          Gene Name:
          midline 1
          Gene Aliases:
          BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY
          Chromosome Location:
          Chr.X: 10445310 - 11111264 on Build GRCh38
          UniGene ID:
          Hs.27695
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS106544
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MID1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000381.3 NP_000372.1 2 423
          NM_001098624.2 NP_001092094.1 2 353
          NM_001193277.1 NP_001180206.1 2 212
          NM_001193278.1 NP_001180207.1 1 78
          NM_001193279.1 NP_001180208.1 1 78
          NM_001193280.1 NP_001180209.1 1 78
          NM_001193281.1 NP_001180210.1 1 78
          NM_033289.1 NP_150631.1 2 423
          NM_033290.3 NP_150632.1 2 215
          XM_005274536.1 XP_005274593.1 2 235
          XM_005274537.1 XP_005274594.1 2 212
          XM_006724492.3 XP_006724555.1 2 834
          XM_011545525.2 XP_011543827.1 4 536
          XM_011545526.2 XP_011543828.1 4 571
          XM_011545527.2 XP_011543829.1 2 481
          XM_017029534.1 XP_016885023.1 2 401
          XM_017029535.1 XP_016885024.1 4 536
          XM_017029536.1 XP_016885025.1 4 651
          XM_017029537.1 XP_016885026.1 4 536
          XM_017029538.1 XP_016885027.1 2 235
          XM_017029539.1 XP_016885028.1 2 397
          XM_017029540.1 XP_016885029.1 4 536
          XM_017029541.1 XP_016885030.1 4 536
          XM_017029542.1 XP_016885031.1 4 536
          GenBank mRNA AF035360.1 - 2 109
          AF041206.1 - 2 303
          AF041208.1 - 2 423
          AF230976.1 - 2 208
          AF230977.1 - 2 208
          AF269101.1 - 3 331
          AK226104.1 - 2 241
          AK291215.1 - 2 305
          AK297205.1 - 2 369
          AK299061.1 - 2 362
          AK315095.1 - 2 254
          AY539988.1 - 1 22
          AY539994.1 - 1 22
          AY540022.1 - 1 22
          AY540023.1 - 1 22
          BC053626.1 - 2 280 6163881
          DA435613.1 - 2 212
          DA856308.1 - 2 215
          DC309977.1 - 2 423
          DC369426.1 - 2 353
          Y13667.1 - 2 208

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          negative regulation of microtubule depolymerization
          pattern specification process
          positive regulation of stress-activated MAPK cascade
          protein localization to microtubule
          interferon-gamma-mediated signaling pathway
          protein binding
          microtubule binding
          zinc ion binding
          ligase activity
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity
          phosphoprotein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix