Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ODF2 SIRNA Assays ›
Gene Symbol:
ODF2
Gene Name:
outer dense fiber of sperm tails 2
Gene Aliases:
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location:
Chr.9: 128455155 - 128501292 on Build GRCh38
UniGene ID:
Hs.129055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS107415
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ODF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242352.1
NP_001229281.1
14
1861
NM_001242353.1
NP_001229282.1
14
1947
NM_001242354.1
NP_001229283.1
13
1633
NM_002540.4
NP_002531.3
16
2149
NM_153432.1
NP_702910.1
13
1707
NM_153433.1
NP_702911.1
14
1874
NM_153435.1
NP_702913.1
14
1767
NM_153436.1
NP_702914.1
13
1531
NM_153437.2
NP_702915.1
14
1797
NM_153439.1
NP_702917.1
13
1650
NM_153440.1
NP_702918.1
13
1577
XM_005252010.2
XP_005252067.1
13
1659
XM_005252011.2
XP_005252068.1
13
1599
XM_005252012.3
XP_005252069.1
14
1849
XM_005252013.3
XP_005252070.1
14
1796
XM_005252014.3
XP_005252071.1
14
1845
XM_005252017.4
XP_005252074.1
14
1750
XM_005252018.2
XP_005252075.1
15
1955
XM_005252019.3
XP_005252076.1
15
1974
XM_005252022.2
XP_005252079.1
14
1773
XM_005252023.2
XP_005252080.1
14
1744
XM_005252024.3
XP_005252081.1
14
1763
XM_005252026.4
XP_005252083.1
14
1692
XM_005252029.2
XP_005252086.1
14
1773
XM_006717126.3
XP_006717189.1
14
1849
XM_011518722.2
XP_011517024.1
14
1773
XM_011518723.2
XP_011517025.1
14
1716
XM_011518724.2
XP_011517026.1
15
1937
XM_011518725.2
XP_011517027.1
15
1941
XM_011518726.2
XP_011517028.1
15
1959
XM_011518728.2
XP_011517030.1
14
1710
XM_011518729.2
XP_011517031.1
14
1646
XM_011518730.2
XP_011517032.1
14
1773
XM_011518731.2
XP_011517033.1
13
1602
XM_011518732.2
XP_011517034.1
14
1559
XM_011518733.2
XP_011517035.1
13
1545
XM_011518744.1
XP_011517046.1
13
1599
XM_017014764.1
XP_016870253.1
15
1930
XM_017014765.1
XP_016870254.1
15
1949
XM_017014766.1
XP_016870255.1
13
1659
XM_017014767.1
XP_016870256.1
15
1941
XM_017014768.1
XP_016870257.1
14
1733
XM_017014769.1
XP_016870258.1
14
1762
XM_017014770.1
XP_016870259.1
15
1917
XM_017014771.1
XP_016870260.1
15
1898
XM_017014772.1
XP_016870261.1
15
1974
XM_017014773.1
XP_016870262.1
15
1950
XM_017014774.1
XP_016870263.1
14
1660
XM_017014775.1
XP_016870264.1
15
1917
XM_017014776.1
XP_016870265.1
15
1898
XM_017014777.1
XP_016870266.1
14
1700
XM_017014778.1
XP_016870267.1
14
1803
XM_017014779.1
XP_016870268.1
13
1544
XM_017014780.1
XP_016870269.1
14
1727
XM_017014781.1
XP_016870270.1
13
1482
GenBank mRNA
AF012549.1
-
14
1737
AF053970.1
-
12
1436
AK093367.1
-
13
1657
AK299303.1
-
14
1709
AK300793.1
-
13
1482
AK301754.1
-
13
1574
AK301842.1
-
13
1499
AK301855.1
-
13
1553
AK302684.1
-
14
1801
AY319414.1
-
13
1683
AY366499.1
-
14
1644
BC010629.1
-
16
1947
3874501
BC091500.1
-
14
1761
30378678
DQ444713.1
-
12
1474
DQ444714.1
-
12
1417
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
multicellular organism development
spermatid development
structural molecule activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0