Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146184.1 | NP_001139656.1 | 9 | 1897 | ||
NM_001146185.1 | NP_001139657.1 | 9 | 1583 | |||
NM_001146186.1 | NP_001139658.1 | 7 | 1930 | |||
NM_001146187.1 | NP_001139659.1 | 9 | 1589 | |||
NM_006210.2 | NP_006201.1 | 10 | 1973 | |||
GenBank mRNA | AB003039.1 | - | 9 | 1546 | ||
AB006625.2 | - | 9 | 1562 | |||
AF208967.1 | - | 10 | 1943 | |||
AF208968.1 | - | 9 | 1867 | |||
AF208969.1 | - | 9 | 1553 | |||
AF208970.1 | - | 7 | 1931 | |||
AK295679.1 | - | 9 | 1589 | |||
AK307476.1 | - | 9 | 1589 | |||
BC052616.1 | - | 6 | 1485 | |||
BC136268.1 | - | 8 | 1417 | 9020255 | ||
BC150272.1 | - | 9 | 1562 | 9007244 |