Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC8A1 SIRNA Assays ›
          Gene Symbol:
          SLC8A1
          Gene Name:
          solute carrier family 8 member A1
          Gene Aliases:
          NCX1
          Chromosome Location:
          Chr.2: 40112146 - 40512452 on Build GRCh38
          UniGene ID:
          Hs.31961
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS109885
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC8A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001112800.1 NP_001106271.1 1 1246
          NM_001112801.1 NP_001106272.1 1 1246
          NM_001112802.1 NP_001106273.1 2 1333
          NM_001252624.1 NP_001239553.1 1 1246
          NM_021097.2 NP_066920.1 1 1246
          XM_005264514.3 XP_005264571.1 2 1428
          XM_006712081.2 XP_006712144.1 2 1428
          XM_006712082.3 XP_006712145.1 3 1530
          XM_006712083.3 XP_006712146.1 2 1347
          XM_006712084.3 XP_006712147.1 3 1628
          XM_006712085.3 XP_006712148.1 2 2250
          XM_011533050.1 XP_011531352.1 1 1243
          XM_011533054.2 XP_011531356.1 2 1264
          XM_011533055.2 XP_011531357.1 2 1391
          XM_011533056.1 XP_011531358.1 2 1264
          XM_011533057.2 XP_011531359.2 2 1412
          XM_011533058.2 XP_011531360.1 2 1263
          XM_017004745.1 XP_016860234.1 2 1412
          XM_017004746.1 XP_016860235.1 2 1265
          XM_017004747.1 XP_016860236.1 2 1467
          XM_017004748.1 XP_016860237.1 2 1333
          XM_017004749.1 XP_016860238.1 3 1533
          XM_017004750.1 XP_016860239.1 4 1767
          XM_017004751.1 XP_016860240.1 4 1764
          XM_017004752.1 XP_016860241.1 3 1568
          XM_017004753.1 XP_016860242.1 2 1264
          XM_017004754.1 XP_016860243.1 2 1392
          XM_017004755.1 XP_016860244.1 2 1391
          XM_017004756.1 XP_016860245.1 2 1471
          XM_017004757.1 XP_016860246.1 2 1466
          XM_017004758.1 XP_016860247.1 2 1448
          XM_017004759.1 XP_016860248.1 2 1454
          XM_017004760.1 XP_016860249.1 2 1428
          XM_017004761.1 XP_016860250.1 2 1471
          XM_017004762.1 XP_016860251.1 2 1446
          XM_017004763.1 XP_016860252.1 2 1428
          XM_017004764.1 XP_016860253.1 2 1263
          GenBank mRNA AF108388.1 - 1 1222
          AF108389.1 - 1 1222
          AF128524.1 - 1 1244
          AK291696.1 - 2 1333
          BX537482.1 - 2 1332
          BX648299.1 - 2 1413
          M91368.1 - 1302
          M96368.1 - 1 1263

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of the force of heart contraction
          regulation of heart rate
          regulation of sodium ion transport
          ion transport
          cellular sodium ion homeostasis
          muscle contraction
          response to nutrient
          response to glucose
          regulation of cell communication by electrical coupling
          positive regulation of fibroblast migration
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          regulation of cardiac muscle contraction by calcium ion signaling
          vascular smooth muscle contraction
          telencephalon development
          positive regulation of bone mineralization
          response to ATP
          cellular response to reactive oxygen species
          sodium ion transmembrane transport
          response to immobilization stress
          response to muscle stretch
          response to drug
          response to hydrogen peroxide
          relaxation of smooth muscle
          negative regulation of cytosolic calcium ion concentration
          cardiac muscle cell development
          calcium ion homeostasis
          relaxation of cardiac muscle
          cardiac muscle contraction
          cytosolic calcium ion transport
          calcium ion transport into cytosol
          calcium ion import
          calcium ion transmembrane transport
          cellular response to caffeine
          cellular response to cAMP
          sodium ion export
          cellular response to hypoxia
          membrane depolarization during cardiac muscle cell action potential
          cell communication by electrical coupling involved in cardiac conduction
          sodium ion import
          positive regulation of the force of heart contraction
          calcium ion export
          regulation of cardiac conduction
          calcium:sodium antiporter activity
          calcium ion binding
          protein binding
          calmodulin binding
          cytoskeletal protein binding
          ankyrin binding
          ion channel binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline