Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167604.1 | NP_001161076.1 | 13 | 1330 | ||
NM_001324128.1 | NP_001311057.1 | 11 | 1177 | |||
NM_001324131.1 | NP_001311060.1 | 13 | 1374 | |||
NM_001324132.1 | NP_001311061.1 | 12 | 1241 | |||
NM_001324133.1 | NP_001311062.1 | 13 | 1330 | |||
NM_001324134.1 | NP_001311063.1 | 11 | 1161 | |||
NM_001324135.1 | NP_001311064.1 | 12 | 1279 | |||
NM_001324136.1 | NP_001311065.1 | 13 | 1545 | |||
NM_020383.3 | NP_065116.3 | 13 | 1330 | |||
XM_011540134.1 | XP_011538436.1 | 13 | 1324 | |||
XM_017016613.1 | XP_016872102.1 | 12 | 1180 | |||
XM_017016614.1 | XP_016872103.1 | 14 | 1313 | |||
XM_017016615.1 | XP_016872104.1 | 12 | 1197 | |||
GenBank mRNA | AF195530.1 | - | 12 | 1157 | ||
AF272981.1 | - | 11 | 1081 | |||
AK093693.1 | - | 11 | 1135 | |||
AK223513.1 | - | 12 | 1141 | |||
AK289436.1 | - | 13 | 1233 | |||
AK295756.1 | - | 5 | 586 | |||
AK304367.1 | - | 13 | 1223 | |||
AL833411.1 | - | 13 | 1236 | |||
BC005126.2 | - | 12 | 1119 | 3836436 | ||
BC007579.1 | - | 13 | 1221 | 3139868 | ||
BC013417.2 | - | 9 | 831 | |||
BX537682.1 | - | 10 | 986 | |||
CR456922.1 | - | 11 | 1081 | |||
X95762.1 | - | 11 | 1081 |