Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF75A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302109.1 | NP_001289038.1 | 5 | 1110 | ||
NM_001324041.1 | NP_001310970.1 | 5 | 1067 | |||
NM_153028.3 | NP_694573.1 | 4 | 586 | |||
XM_011522646.2 | XP_011520948.1 | 5 | 1110 | |||
XM_011522647.2 | XP_011520949.1 | 4 | 684 | |||
XM_017023642.1 | XP_016879131.1 | 4 | 482 | |||
XM_017023643.1 | XP_016879132.1 | 5 | 1110 | |||
XM_017023644.1 | XP_016879133.1 | 5 | 1110 | |||
GenBank mRNA | AK056091.1 | - | 4 | 509 | ||
BC119652.1 | - | 3 | 311 | 40115880 | ||
BC119653.1 | - | 3 | 311 | 40115881 | ||
CR749782.1 | - | 4 | 489 |