Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142401.1 | NP_001135873.1 | 1 | 294 | ||
NM_001142402.1 | NP_001135874.1 | 1 | 294 | |||
NM_001142403.1 | NP_001135875.1 | 1 | 294 | |||
NM_001142404.1 | NP_001135876.1 | 1 | 294 | |||
NM_006016.4 | NP_006007.2 | 1 | 294 | |||
GenBank mRNA | AF106518.1 | - | 1 | 113 | ||
AF263279.1 | - | 1 | 113 | |||
AF299341.1 | - | 1 | 113 | |||
AF299342.1 | - | 1 | 191 | |||
AF299343.1 | - | 1 | 191 | |||
AK298684.1 | - | 1 | 146 | |||
AK301692.1 | - | 1 | 204 | |||
AK303525.1 | - | 1 | 146 | |||
AK307193.1 | - | 1 | 202 | |||
AK312357.1 | - | 1 | 202 | |||
AK315908.1 | - | 1 | 145 | |||
BC011522.2 | - | 1 | 194 | 3854912 | ||
D14043.1 | - | 1 | 192 | |||
DC403839.1 | - | 1 | 294 | |||
FJ200494.1 | - | 1 | 113 |