Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PIAS2 SIRNA Assays ›
          Gene Symbol:
          PIAS2
          Gene Name:
          protein inhibitor of activated STAT 2
          Gene Aliases:
          ARIP3, DIP, MIZ, MIZ1, PIASX, PIASX-ALPHA, PIASX-BETA, SIZ2, ZMIZ4
          Chromosome Location:
          Chr.18: 46803224 - 46920167 on Build GRCh38
          UniGene ID:
          Hs.57769
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS113395
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PIAS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001324046.1 NP_001310975.1 8, 9 1237
          NM_001324047.1 NP_001310976.1 7, 8 1148
          NM_001324048.1 NP_001310977.1 8, 9 1237
          NM_001324049.1 NP_001310978.1 9, 10 1360
          NM_001324051.1 NP_001310980.1 7, 8 1148
          NM_001324052.1 NP_001310981.1 9, 10 1338
          NM_001324053.1 NP_001310982.1 9, 10 1497
          NM_001324054.1 NP_001310983.1 7, 8 1128
          NM_001324055.1 NP_001310984.1 8, 9 1256
          NM_001324057.1 NP_001310986.1 9, 10 1338
          NM_001324058.1 NP_001310987.1 6, 7 1039
          NM_001324059.1 NP_001310988.1 8, 9 1237
          NM_001324060.1 NP_001310989.1 6, 7 937
          NM_004671.4 NP_004662.2 6, 7 1039
          NM_173206.3 NP_775298.1 6, 7 1039
          NR_136684.1 5, 6 954
          NR_136685.1 5, 6 564
          XM_005258377.3 XP_005258434.1 6, 7 937
          XM_005258379.3 XP_005258436.1 6, 7 1054
          XM_006722571.3 XP_006722634.1 7, 8 1022
          XM_006722572.3 XP_006722635.1 7, 8 1022
          XM_006722573.2 XP_006722636.1 7, 8 1022
          XM_011526258.2 XP_011524560.1 6, 7 1102
          XM_011526259.2 XP_011524561.1 7, 8 953
          XM_017026068.1 XP_016881557.1 9, 10 1338
          XM_017026069.1 XP_016881558.1 7, 8 1071
          XM_017026070.1 XP_016881559.1 9, 10 1338
          XM_017026071.1 XP_016881560.1 7, 8 953
          XM_017026072.1 XP_016881561.1 6, 7 892
          GenBank mRNA AF077953.1 - 6, 7 852
          AF077954.1 - 6, 7 852
          AF361054.1 - 6, 7 930
          AK294801.1 - 5, 6 537
          AK308143.1 - 5, 6 876
          BC015190.1 - 6, 7 1010 4080297
          BC111060.1 - 6, 7 981 5745101

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          protein sumoylation
          androgen receptor signaling pathway
          regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of androgen receptor signaling pathway
          DNA binding
          transcription coactivator activity
          protein binding
          transcription factor binding
          zinc ion binding
          ligase activity
          SUMO transferase activity
          ubiquitin protein ligase binding
          androgen receptor binding
          SUMO ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix