Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DOCK9 SIRNA Assays ›
          Gene Symbol:
          DOCK9
          Gene Name:
          dedicator of cytokinesis 9
          Gene Aliases:
          ZIZ1, ZIZIMIN1
          Chromosome Location:
          Chr.13: 98793487 - 99086406 on Build GRCh38
          UniGene ID:
          Hs.596105
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS118628
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DOCK9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130048.1 NP_001123520.1 27, 28 3115
          NM_001130049.1 NP_001123521.1 27, 28 2998
          NM_001130050.1 NP_001123522.1 27, 28 3115
          NM_001318849.1 NP_001305778.1 27, 28 2998
          NM_015296.2 NP_056111.1 27, 28 2998
          XM_005254034.3 XP_005254091.1 27, 28 2987
          XM_005254035.3 XP_005254092.1 27, 28 2987
          XM_006719922.1 XP_006719985.1 27, 28 3148
          XM_006719924.2 XP_006719987.1 27, 28 2987
          XM_006719925.1 XP_006719988.1 27, 28 3115
          XM_006719931.3 XP_006719994.1 27, 28 2943
          XM_006719932.1 XP_006719995.1 27, 28 3115
          XM_006719936.1 XP_006719999.1 27, 28 3148
          XM_006719937.3 XP_006720000.1 27, 28 2943
          XM_017020503.1 XP_016875992.1 27, 28 2988
          XM_017020504.1 XP_016875993.1 27, 28 2988
          XM_017020505.1 XP_016875994.1 27, 28 2988
          XM_017020506.1 XP_016875995.1 27, 28 2988
          XM_017020507.1 XP_016875996.1 27, 28 2988
          XM_017020508.1 XP_016875997.1 27, 28 2988
          XM_017020509.1 XP_016875998.1 27, 28 3115
          XM_017020510.1 XP_016875999.1 27, 28 2988
          XM_017020511.1 XP_016876000.1 27, 28 2988
          XM_017020512.1 XP_016876001.1 27, 28 3115
          XM_017020513.1 XP_016876002.1 27, 28 2988
          XM_017020514.1 XP_016876003.1 27, 28 3115
          XM_017020515.1 XP_016876004.1 27, 28 3115
          XM_017020516.1 XP_016876005.1 27, 28 3115
          XM_017020517.1 XP_016876006.1 27, 28 3087
          XM_017020518.1 XP_016876007.1 27, 28 3087
          XM_017020519.1 XP_016876008.1 27, 28 3115
          XM_017020520.1 XP_016876009.1 27, 28 3115
          XM_017020521.1 XP_016876010.1 27, 28 3148
          GenBank mRNA AB028981.2 - 27, 28 3021
          AF527605.1 - 27, 28 2998
          AK122987.1 - 16, 17 1602
          AK126492.1 - 27, 28 3115
          AK127329.1 - 27, 28 3087
          AK127508.1 - 25, 26 2697
          BC136274.1 - 27, 28 2978 9020261
          BC146814.1 - 27, 28 2992 8860392
          BC171753.1 - 27, 28 2978 9053254
          CR936820.1 - 27, 28 3078

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          blood coagulation
          biological_process
          positive regulation of GTPase activity
          cell-cell adhesion
          guanyl-nucleotide exchange factor activity
          protein binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-867f8cf58d-sfwzd:80/100.66.130.62:80.
          git-commit: 7a7e1ef32845d1da800a461bcf18a29ea3e3afc0
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix