Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM149A SIRNA Assays ›
          Gene Symbol:
          FAM149A
          Gene Name:
          family with sequence similarity 149 member A
          Gene Aliases:
          MST119, MSTP119
          Chromosome Location:
          Chr.4: 186104798 - 186172663 on Build GRCh38
          UniGene ID:
          Hs.744961
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS119404
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM149A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006655.2 NP_001006656.1 7, 8 1016
          NM_015398.2 NP_056213.1 8, 9 1315
          XM_005262911.1 XP_005262968.1 8, 9 1276
          XM_005262912.1 XP_005262969.1 8, 9 1347
          XM_005262913.1 XP_005262970.1 8, 9 1184
          XM_011531834.1 XP_011530136.1 7, 8 1043
          XM_011531835.2 XP_011530137.1 8, 9 1276
          XM_011531836.1 XP_011530138.1 7, 8 1043
          XM_017007996.1 XP_016863485.1 8, 9 1861
          XM_017007997.1 XP_016863486.1 8, 9 1858
          XM_017007998.1 XP_016863487.1 7, 8 1043
          XM_017007999.1 XP_016863488.1 8, 9 2648
          XM_017008000.1 XP_016863489.1 8, 9 1132
          XM_017008001.1 XP_016863490.1 8, 9 1221
          XM_017008002.1 XP_016863491.1 8, 9 1276
          XM_017008003.1 XP_016863492.1 8, 9 1132
          XM_017008004.1 XP_016863493.1 8, 9 1184
          XM_017008005.1 XP_016863494.1 8, 9 1289
          XM_017008006.1 XP_016863495.1 8, 9 1184
          XM_017008007.1 XP_016863496.1 8, 9 1347
          GenBank mRNA AF173894.1 - 7, 8 951
          AK057166.1 - 8, 9 1145
          AK297679.1 - 8, 9 1255
          AL080065.1 - 2, 3 364
          BC110816.1 - 8, 9 1217
          BC136792.1 - 5, 6 761 9020782
          BC142992.1 - 8, 9 1269 8860446
          BC144411.1 - 5, 6 761 9052932

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix