Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013382.5 | NP_037514.2 | 19 | 2142 | ||
XM_011536675.2 | XP_011534977.1 | 20 | 2331 | |||
XM_011536676.2 | XP_011534978.1 | 19 | 2244 | |||
XM_011536677.2 | XP_011534979.1 | 16 | 1873 | |||
XM_011536679.2 | XP_011534981.1 | 16 | 1533 | |||
XM_017021244.1 | XP_016876733.1 | 18 | 1976 | |||
XM_017021245.1 | XP_016876734.1 | 15 | 1430 | |||
GenBank mRNA | AB209114.1 | - | 18 | 2917 | ||
AF105020.1 | - | 19 | 2145 | |||
AK023804.1 | - | 13 | 2733 | |||
AK025962.1 | - | 9 | 761 | |||
AL353956.1 | - | 2 | 108 | |||
AY090480.1 | - | 19 | 2145 | |||
BC031651.1 | - | 19 | 2032 | 5169145 | ||
BE797202.1 | - | 547 | ||||
CR627447.1 | - | 19 | 1712 | |||
DA056935.1 | - | 4 | 299 |