Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015987.4 | NP_057071.2 | 4 | 694 | ||
GenBank mRNA | AF117615.1 | - | 4 | 577 | ||
AF167473.1 | - | 4 | 499 | |||
AK289877.1 | - | 4 | 658 | |||
BC016277.2 | - | 4 | 529 | 3907035 | ||
BT007294.1 | - | 4 | 499 | |||
CF126276.1 | - | 4 | 695 |
Gene Symbol: LOC100132368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131565.1 | - | 1 | 1971 |