Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291822.1 | NP_001278751.1 | 1 | 77 | ||
NM_001291823.1 | NP_001278752.1 | 1 | 77 | |||
NM_016523.2 | NP_057607.1 | 1 | 77 | |||
NR_120305.1 | 1 | 77 | ||||
XM_017019414.1 | XP_016874903.1 | 1 | 196 | |||
XM_017019415.1 | XP_016874904.1 | 1 | 255 | |||
GenBank mRNA | AF175206.1 | - | 1 | 77 | ||
AF175207.1 | - | 1 | 77 | |||
AF267244.1 | - | 1 | 77 | |||
AF267245.1 | - | 1 | 77 | |||
AJ305370.1 | - | 1 | 60 | |||
BC096734.1 | - | 1 | 37 | |||
BC098166.1 | - | 1 | 37 | 40015535 | ||
BC098257.1 | - | 1 | 37 | 40015534 | ||
BC098354.1 | - | 1 | 37 | 40015531 |