Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM49B SIRNA Assays ›
Gene Symbol:
FAM49B
Gene Name:
family with sequence similarity 49 member B
Gene Aliases:
BM-009, L1
Chromosome Location:
Chr.8: 129839593 - 130016697 on Build GRCh38
UniGene ID:
Hs.126941
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS122372
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM49B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256763.1
NP_001243692.1
5
493
NM_016623.4
NP_057707.3
4
406
NR_046359.1
7
759
NR_046360.1
7
669
NR_046361.1
6
659
XM_005250966.3
XP_005251023.1
3
262
XM_011517108.2
XP_011515410.1
6
555
XM_011517109.2
XP_011515411.1
6
327
XM_011517110.2
XP_011515412.1
5
467
XM_011517111.2
XP_011515413.1
5
285
XM_011517112.2
XP_011515414.1
7
510
XM_011517113.2
XP_011515415.1
6
521
XM_011517114.2
XP_011515416.1
6
427
XM_011517115.2
XP_011515417.1
5
434
XM_011517116.2
XP_011515418.1
6
468
XM_011517117.2
XP_011515419.1
5
437
XM_011517118.2
XP_011515420.1
5
381
XM_011517120.2
XP_011515422.1
4
380
XM_011517121.2
XP_011515423.1
7
392
XM_011517122.2
XP_011515424.1
5
471
XM_011517123.2
XP_011515425.1
5
244
XM_011517124.2
XP_011515426.1
4
359
XM_011517125.2
XP_011515427.1
4
298
XM_017013539.1
XP_016869028.1
7
514
XM_017013540.1
XP_016869029.1
7
373
XM_017013541.1
XP_016869030.1
5
501
XM_017013542.1
XP_016869031.1
7
551
XM_017013543.1
XP_016869032.1
7
651
XM_017013544.1
XP_016869033.1
7
433
XM_017013545.1
XP_016869034.1
8
697
XM_017013546.1
XP_016869035.1
6
464
XM_017013547.1
XP_016869036.1
8
556
XM_017013548.1
XP_016869037.1
6
560
XM_017013549.1
XP_016869038.1
6
423
XM_017013550.1
XP_016869039.1
5
474
XM_017013551.1
XP_016869040.1
7
469
XM_017013552.1
XP_016869041.1
7
514
XM_017013553.1
XP_016869042.1
6
327
XM_017013554.1
XP_016869043.1
8
539
XM_017013555.1
XP_016869044.1
5
494
XM_017013556.1
XP_016869045.1
5
240
XM_017013557.1
XP_016869046.1
6
286
XM_017013558.1
XP_016869047.1
6
431
XM_017013559.1
XP_016869048.1
6
290
XM_017013560.1
XP_016869049.1
4
405
XM_017013561.1
XP_016869050.1
4
157
XM_017013562.1
XP_016869051.1
5
344
GenBank mRNA
AF208851.1
-
6
344
AK002059.1
-
7
469
AK130372.1
-
4
293
AY598320.1
-
2
92
BC003599.1
-
5
272
3501828
BC016345.1
-
6
413
4041187
BC017297.2
-
7
641
5017493
BX419336.2
-
5
493
CR749628.1
-
5
239
DA371541.1
-
5
476
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0