Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST7L SIRNA Assays ›
Gene Symbol:
ST7L
Gene Name:
suppression of tumorigenicity 7 like
Gene Aliases:
FAM4B, ST7R, STLR
Chromosome Location:
Chr.1: 112517444 - 112620838 on Build GRCh38
UniGene ID:
Hs.201921
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS123438
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST7L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308264.1
NP_001295193.1
11
1452
NM_017744.4
NP_060214.2
12
1661
NM_138727.3
NP_620055.1
12
1610
NM_138728.2
NP_620056.1
12
1661
NM_138729.3
NP_620057.1
12
1661
XM_005270963.4
XP_005271020.1
12
1184
XM_005270964.3
XP_005271021.1
11
1276
XM_011541627.2
XP_011539929.1
12
1370
XM_011541628.2
XP_011539930.1
12
1370
XM_011541629.2
XP_011539931.1
12
1384
XM_011541630.2
XP_011539932.1
12
2080
XM_011541631.2
XP_011539933.1
12
1208
XM_017001534.1
XP_016857023.1
12
1370
XM_017001536.1
XP_016857025.1
11
1276
XM_017001537.1
XP_016857026.1
10
1096
XM_017001538.1
XP_016857027.1
10
996
GenBank mRNA
AB081317.1
-
12
1432
AB081318.1
-
12
1381
AB081319.1
-
12
1432
AB081320.1
-
12
1432
AK097837.1
-
11
1241
AK295955.1
-
10
1164
AK301869.1
-
12
1362
AK308727.1
-
11
1324
BC027898.1
-
10
1148
BC037830.1
-
13
2151
4812133
BC047392.1
-
12
1387
5273549
BC065501.1
-
12
1568
4417786
BX537988.1
-
13
1523
BX538338.1
-
7
685
BX640667.1
-
12
1337
Back To Top
More Information
Gene Ontology Categories:
Function(s)
negative regulation of cell growth
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0