Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWF19L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303404.1 | NP_001290333.1 | 11 | 1321 | ||
NM_001303405.1 | NP_001290334.1 | 11 | 1366 | |||
NM_001303406.1 | NP_001290335.1 | 8 | 1055 | |||
NM_001303407.1 | NP_001290336.1 | 10 | 1090 | |||
NM_018294.5 | NP_060764.3 | 11 | 1321 | |||
GenBank mRNA | AK000876.1 | - | 1 | 161 | ||
AK001860.1 | - | 11 | 1267 | |||
AK023984.1 | - | 9 | 918 | |||
AK055313.1 | - | 2 | 1387 | |||
AK097895.1 | - | 10 | 1089 | |||
AK225590.1 | - | 11 | 1279 | |||
AK295303.1 | - | 8 | 1025 | |||
AL832515.1 | - | 11 | 1278 | |||
BC008746.2 | - | 11 | 1262 | 3345869 | ||
KU178692.1 | - | 11 | 1233 | |||
KU178693.1 | - | 11 | 1217 |