Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNF331 SIRNA Assays ›
          Gene Symbol:
          ZNF331
          Gene Name:
          zinc finger protein 331
          Gene Aliases:
          RITA, ZNF361, ZNF463
          Chromosome Location:
          Chr.19: 53503819 - 53580269 on Build GRCh38
          UniGene ID:
          Hs.185674
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS124486
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNF331
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001079906.1 NP_001073375.1 6 783
          NM_001079907.1 NP_001073376.1 6 803
          NM_001253798.1 NP_001240727.1 7 801
          NM_001253799.1 NP_001240728.1 6 771
          NM_001253800.1 NP_001240729.1 6 730
          NM_001253801.2 NP_001240730.1 5 693
          NM_001317113.1 NP_001304042.1 5 719
          NM_001317114.1 NP_001304043.1 6 636
          NM_001317115.1 NP_001304044.1 7 813
          NM_001317116.1 NP_001304045.1 5 774
          NM_001317117.1 NP_001304046.1 6 723
          NM_001317118.1 NP_001304047.1 6 804
          NM_001317119.1 NP_001304048.1 5 773
          NM_001317120.1 NP_001304049.1 7 1757
          NM_001317121.1 NP_001304050.1 5 677
          NM_018555.5 NP_061025.5 7 1795
          XM_011527076.2 XP_011525378.1 8 2314
          XM_011527078.2 XP_011525380.1 8 1913
          XM_017026936.1 XP_016882425.1 12 2507
          XM_017026937.1 XP_016882426.1 12 2032
          XM_017026938.1 XP_016882427.1 7 1026
          XM_017026939.1 XP_016882428.1 6 798
          XM_017026940.1 XP_016882429.1 5 678
          GenBank mRNA AF251515.2 - 6 737
          AF272148.1 - 5 547
          AK075226.1 - 6 704
          AK128736.1 - 7 1681
          AK312889.1 - 5 655
          BC009365.2 - 5 520
          BC009433.1 - 8 1020 3355780
          CD639405.1 - 6 593
          CR749560.1 - 7 1705

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc