Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IFT122 SIRNA Assays ›
          Gene Symbol:
          IFT122
          Gene Name:
          intraflagellar transport 122
          Gene Aliases:
          CED, CED1, SPG, WDR10, WDR10p, WDR140
          Chromosome Location:
          Chr.3: 129440036 - 129520507 on Build GRCh38
          UniGene ID:
          Hs.655284
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS124989
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IFT122
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001280541.1 NP_001267470.1 9, 10 1096
          NM_001280545.1 NP_001267474.1 10, 11 1271
          NM_001280546.1 NP_001267475.1 7, 8 864
          NM_018262.3 NP_060732.2 8, 9 943
          NM_052985.3 NP_443711.2 10, 11 1273
          NM_052989.2 NP_443715.1 9, 10 1120
          NM_052990.2 NP_443716.1 6, 7 787
          XM_005247609.1 XP_005247666.1 8, 9 908
          XM_006713689.1 XP_006713752.1 9, 10 1085
          XM_006713691.2 XP_006713754.1 9, 10 1085
          XM_006713692.2 XP_006713755.1 8, 9 908
          XM_006713695.2 XP_006713758.1 9, 10 1085
          XM_011512972.2 XP_011511274.1 9, 10 1085
          XM_017006817.1 XP_016862306.1 9, 10 1085
          XM_017006818.1 XP_016862307.1 9, 10 1085
          XM_017006819.1 XP_016862308.1 9, 10 1085
          XM_017006820.1 XP_016862309.1 9, 10 1085
          XM_017006821.1 XP_016862310.1 9, 10 1085
          XM_017006822.1 XP_016862311.1 7, 8 876
          XM_017006823.1 XP_016862312.1 8, 9 908
          XM_017006824.1 XP_016862313.1 8, 9 908
          XM_017006825.1 XP_016862314.1 8, 9 908
          XM_017006826.1 XP_016862315.1 8, 9 908
          XM_017006827.1 XP_016862316.1 8, 9 908
          XM_017006828.1 XP_016862317.1 6, 7 699
          XM_017006829.1 XP_016862318.1 6, 7 699
          XM_017006830.1 XP_016862319.1 10, 11 1083
          XM_017006831.1 XP_016862320.1 8, 9 1004
          XM_017006832.1 XP_016862321.1 9, 10 983
          XM_017006833.1 XP_016862322.1 6, 7 699
          XM_017006834.1 XP_016862323.1 6, 7 699
          XM_017006835.1 XP_016862324.1 7, 8 827
          XM_017006836.1 XP_016862325.1 9, 10 906
          XM_017006838.1 XP_016862327.1 5, 6 450
          GenBank mRNA AF244931.1 - 10, 11 1170
          AF302154.1 - 9, 10 902
          AK001759.1 - 8, 9 756
          AK022525.1 - 8, 9 854
          AK024435.1 - 7, 8 631
          AK122965.1 - 3, 4 738
          AK124140.1 - 10, 11 1074
          AK223095.1 - 8, 9 755
          AK293852.1 - 6, 7 601
          AK294515.1 - 4, 5 492
          AK298526.1 - 5, 6 364
          AK299903.1 - 7, 8 577
          AK315103.1 - 9, 10 920
          AL133565.1 - 7, 8 697
          BC003045.1 - 6, 7 579 2823536
          BC004238.2 - 6, 7 579 2823536
          BC028353.2 - 9, 10 860 4836844

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          signal transduction downstream of smoothened
          embryonic body morphogenesis
          negative regulation of smoothened signaling pathway involved in ventral spinal cord patterning
          embryonic heart tube development
          embryonic forelimb morphogenesis
          intraciliary anterograde transport
          intraciliary retrograde transport
          intraciliary transport involved in cilium morphogenesis
          cilium assembly
          embryonic digit morphogenesis
          negative regulation of smoothened signaling pathway
          camera-type eye morphogenesis
          negative regulation of epithelial cell proliferation
          limb development
          cilium morphogenesis
          ciliary receptor clustering involved in smoothened signaling pathway
          smoothened signaling pathway involved in dorsal/ventral neural tube patterning
          embryonic heart tube left/right pattern formation
          protein localization to cilium
          establishment of protein localization to organelle
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0