Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CEACAM19 SIRNA Assays ›
          Gene Symbol:
          CEACAM19
          Gene Name:
          carcinoembryonic antigen related cell adhesion molecule 19
          Gene Aliases:
          CEACM19, CEAL1
          Chromosome Location:
          Chr.19: 44671452 - 44684355 on Build GRCh38
          UniGene ID:
          Hs.416925
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS125838
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CEACAM19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127893.1 NP_001121365.1 3 970
          NM_020219.3 NP_064604.2 3 970
          GenBank mRNA AK027157.1 - 3 687
          AK291588.1 - 3 671
          AY358394.1 - 3 700
          BC083499.1 - 3 970 30390224
          KJ851726.1 - 3 490
          KJ851727.1 - 3 490
          KJ851728.1 - 3 490
          KJ851729.1 - 3 490
          KJ851730.1 - 3 490
          KJ851731.1 - 3 490
          KJ851732.1 - 3 490
          KJ851733.1 - 3 490
          KJ851734.1 - 3 490
          KJ851735.1 - 3 490
          KJ851736.1 - 3 490
          KJ851737.1 - 3 490
          KJ851738.1 - 3 490
          KJ851739.1 - 3 490
          KJ851740.1 - 3 490
          KJ851741.1 - 3 490
          KJ851760.1 - 4 624
          KJ851761.1 - 4 624
          KJ851762.1 - 4 624
          KJ851763.1 - 4 624
          KJ851764.1 - 4 624
          KJ851765.1 - 4 624
          KJ851766.1 - 4 624
          KJ851767.1 - 4 624
          KJ851768.1 - 4 624
          KJ851769.1 - 4 624
          KJ851770.1 - 4 624
          KJ851771.1 - 4 624
          KJ851772.1 - 4 624
          KJ851773.1 - 4 624
          KJ851774.1 - 4 624
          KJ851775.1 - 4 624
          KJ851776.1 - 4 624
          KJ851777.1 - 4 624
          KJ851796.1 - 3 490
          KJ851797.1 - 4 624

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix