Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020247.4 | NP_064632.2 | 12 | 1647 | ||
XM_005273201.1 | XP_005273258.1 | 12 | 1543 | |||
XM_011544238.1 | XP_011542540.1 | 13 | 1739 | |||
XM_011544239.2 | XP_011542541.1 | 14 | 1762 | |||
XM_011544240.2 | XP_011542542.1 | 12 | 1556 | |||
XM_011544241.2 | XP_011542543.1 | 12 | 1518 | |||
XM_017001852.1 | XP_016857341.1 | 12 | 1737 | |||
GenBank mRNA | AB073905.1 | - | 12 | 1570 | ||
AF218003.1 | - | 4 | 662 | |||
AJ278126.1 | - | 8 | 865 | |||
AK074693.1 | - | 12 | 1590 | |||
AK090494.1 | - | 12 | 1384 | |||
AK092784.1 | - | 12 | 1370 | |||
AK126200.1 | - | 5 | 2335 | |||
AK293571.1 | - | 9 | 1010 | |||
AK297735.1 | - | 9 | 926 | |||
AK300537.1 | - | 8 | 736 | |||
AK301012.1 | - | 14 | 1568 | |||
AL137511.1 | - | 4 | 1189 | |||
BC005171.2 | - | 12 | 1596 | 3604948 | ||
BX648860.1 | - | 12 | 1468 |