Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIDINS220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020738.2 | NP_065789.1 | 25 | 3618 | ||
GenBank mRNA | AB033076.2 | - | 25 | 3593 | ||
AK074254.1 | - | 5 | 514 | |||
AK294464.1 | - | 2 | 250 | |||
BX640878.1 | - | 2 | 174 | |||
KJ812119.1 | - | 2 | 244 | |||
KJ812120.1 | - | 2 | 244 |