Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020752.2 | NP_065803.2 | 7 | 1644 | ||
XM_017016452.1 | XP_016871941.1 | 4 | 464 | |||
GenBank mRNA | AK095120.1 | - | 7 | 1942 | ||
AY528411.1 | - | 7 | 1644 |